missing translation for 'onlineSavingsMsg'
Learn More

Allyl Methyl Sulfide 97.0+%, TCI America™

Catalog No. p-7117251
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11175ML 5 mL
A111725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A11175ML Supplier TCI America Supplier No. A11175ML
Only null left

Chemical Identifiers

CAS 10152-76-8
Molecular Formula C4H8S
Molecular Weight (g/mol) 88.17
MDL Number MFCD00008657
InChI Key NVLPQIPTCCLBEU-UHFFFAOYSA-N
Synonym allyl methyl sulfide, 1-propene, 3-methylthio, allyl methyl sulphide, 3-methylthio-1-propene, sulfide, allyl methyl, methyl propenyl sulfide, unii-v7qi1r316c, methyl allyl sulfide, ccris 7066
PubChem CID 66282
IUPAC Name 3-(methylsulfanyl)prop-1-ene
SMILES CSCC=C

Specifications

Color Colorless
Boiling Point 92°C
Quantity 5 mL
UN Number 1993
Formula Weight 88.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Methyl Sulfide
EINECSNumber (2)-0468
RTECSNumber UD1015000
TSCA Yes