missing translation for 'onlineSavingsMsg'
Learn More

Allyl Methyl Ether 97.0+%, TCI America™

Catalog No. A19835G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A19835G Supplier TCI America Supplier No. A19835G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 627-40-7
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00008649
InChI Key FASUFOTUSHAIHG-UHFFFAOYSA-N
Synonym allyl methyl ether, 1-propene, 3-methoxy, 3-methoxy-1-propene, ether, allyl methyl, 3-methoxypropene, methyl allyl ether, allylmethylether, 4-oxapent-1-ene, 1-methoxy-2-propene, methylallyl ether
PubChem CID 69392
IUPAC Name 3-methoxyprop-1-ene
SMILES COCC=C

Specifications

Color Yellow
Boiling Point 43°C
Quantity 5 g
UN Number 3271
Formula Weight 72.11
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Methyl Ether
TSCA Yes
Recommended Storage Refrigerator