accessibility menu, dialog, popup

UserName

CAS RN 627-40-7

IUPAC Name: 3-methoxyprop-1-ene
Synonyms: methallyl alcohol
Molecular Weight (g/mol): 72.11
Molecular Formula: C4H8O
InChi Key: FASUFOTUSHAIHG-UHFFFAOYSA-N
SMILES: COCC=C
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)