missing translation for 'onlineSavingsMsg'
Learn More

Allyl methyl ether, 94%

Catalog No. p-7049687
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1865209 10 g
AA1865218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1865209 Supplier Thermo Scientific Chemicals Supplier No. 01865209
Only null left

CAS: 627-40-7 | C4H8O | 72.11 g/mol

Allyl methyl ether it used as pharmaceutical intermediate. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 627-40-7
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00008649
InChI Key FASUFOTUSHAIHG-UHFFFAOYSA-N
Synonym allyl methyl ether, 1-propene, 3-methoxy, 3-methoxy-1-propene, ether, allyl methyl, 3-methoxypropene, methyl allyl ether, allylmethylether, 4-oxapent-1-ene, 1-methoxy-2-propene, methylallyl ether
PubChem CID 69392
IUPAC Name 3-methoxyprop-1-ene
SMILES COCC=C

Specifications

Density 0.77 g/mL
Boiling Point 42°C to 43°C
Flash Point -23°C (-94°F)
Odor Ether-like
Refractive Index 1.3803
Quantity 10 g
UN Number UN3271
Formula Weight 72.1
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material Allyl methyl ether
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 210-997-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only