Learn More
Allyl isothiocyanate, 94%, stab.
CAS: 57-06-7 | C4H5NS | 99.151 g/mol
Supplier: Thermo Scientific Chemicals L0290118
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Allyl isothiocyanate | |
| 57-06-7 | |
| 1.013 | |
| 46°C (114°F) | |
| C4H5NS | |
| MFCD00004822 | |
| UN1545 | |
| Moisture sensitive | |
| allyl isothiocyanate, mustard oil, redskin, allylsenevol, allylsenfoel, allyl mustard oil, oleum sinapis, oil of mustard, allylsevenolum, senfoel | |
| C=CCN=C=S | |
| 99.151 | |
| CHEBI:73224 | |
| 94% |
| stabilized | |
| -80°C | |
| 149°C to 151°C | |
| Pungent | |
| 1.53 | |
| 50 g | |
| 773748 | |
| 14,295 | |
| ZOJBYZNEUISWFT-UHFFFAOYSA-N | |
| 3-isothiocyanatoprop-1-ene | |
| 5971 | |
| 99.16 |
Chemical Identifiers
| 57-06-7 | |
| 99.151 | |
| ZOJBYZNEUISWFT-UHFFFAOYSA-N | |
| 5971 | |
| 3-isothiocyanatoprop-1-ene |
| C4H5NS | |
| MFCD00004822 | |
| allyl isothiocyanate, mustard oil, redskin, allylsenevol, allylsenfoel, allyl mustard oil, oleum sinapis, oil of mustard, allylsevenolum, senfoel | |
| CHEBI:73224 | |
| C=CCN=C=S |
Safety and Handling
GHS H Statement
H301-H310-H331-H318-H226-H315-H335
Toxic if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Causes serious eye damage.
Flammable liquid and vapor.
Causes skin irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H301-H310+H330-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ALLYL ISOTHIOCYANATE, STABILIZED
EINECSNumber : 200-309-2
RTECSNumber : NX8225000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only