missing translation for 'onlineSavingsMsg'
Learn More

Allyl isothiocyanate, 94%, stab.

Catalog No. AAL0290106
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0290106 5 g
AAL0290118 50 g
AAL0290130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0290106 Supplier Thermo Scientific Chemicals Supplier No. L0290106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 57-06-7 | C4H5NS | 99.151 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57-06-7
Molecular Formula C4H5NS
Molecular Weight (g/mol) 99.151
MDL Number MFCD00004822
InChI Key ZOJBYZNEUISWFT-UHFFFAOYSA-N
Synonym allyl isothiocyanate, mustard oil, redskin, allylsenevol, allylsenfoel, allyl mustard oil, oleum sinapis, oil of mustard, allylsevenolum, senfoel
PubChem CID 5971
ChEBI CHEBI:73224
IUPAC Name 3-isothiocyanatoprop-1-ene
SMILES C=CCN=C=S

Specifications

Melting Point -80°C
Density 1.013
Boiling Point 149°C to 151°C
Flash Point 46°C (114°F)
Odor Pungent
Refractive Index 1.53
Quantity 5 g
UN Number UN1545
Beilstein 773748
Sensitivity Moisture sensitive
Merck Index 14,295
Formula Weight 99.16
Percent Purity 94%
Chemical Name or Material Allyl isothiocyanate, stabilized
Show More Show Less
Hazard Category H226-H301-H310+H330-H315-H319-H335
Hazard Statement GHS H Statement
H301-H310-H331-H318-H226-H315-H335
Toxic if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Causes serious eye damage.
Flammable liquid and vapor.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ALLYL ISOTHIOCYANATE, STABILIZED
EINECSNumber 200-309-2
RTECSNumber NX8225000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only