missing translation for 'onlineSavingsMsg'
Learn More

Allyl iodide, 97+%, stab. with copper, Thermo Scientific Chemicals

Catalog No. p-7081727
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1182218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1182218 Supplier Thermo Scientific Chemicals Supplier No. A1182218

Allyl iodide is used as a reagent used with allylindium sesquiiodide in the cis-double allylation of cyclopropenes. It is used in synthesis of other organic compounds such as N-alkyl 2-pyrrolidone, Sorbic acid ester etc,.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyl iodide is used as a reagent used with allylindium sesquiiodide in the cis-double allylation of cyclopropenes. It is used in synthesis of other organic compounds such as N-alkyl 2-pyrrolidone, Sorbic acid ester etc,.

Solubility
Soluble in alcohol, chloroform, ether, and diethyl ether. Insoluble in water.

Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents. Incompatible with heat, light, amines, nitrides, azo/diazo compounds, alkali metals, and epoxides.

Chemical Identifiers

CAS 556-56-9
Molecular Formula C3H5I
Molecular Weight (g/mol) 167.98
MDL Number MFCD00001094
InChI Key HFEHLDPGIKPNKL-UHFFFAOYSA-N
Synonym allyl iodide, 1-propene, 3-iodo, 3-iodopropene, 3-iodo-1-propene, 3-iodopropylene, propene, 3-iodo, allyliodide, unii-46830qoa4d, 3-iodo-propene, 3-iodanylprop-1-ene
PubChem CID 11166
IUPAC Name 3-iodoprop-1-ene
SMILES ICC=C

Specifications

Melting Point -99°C
Density 1.832
Boiling Point 101°C to 103°C
Flash Point 18°C (64°F)
Refractive Index 1.554
Linear Formula CH2=CHCH2I
Quantity 50 g
UN Number UN1723
Beilstein 1697594
Sensitivity Light sensitive
Merck Index 14,294
Solubility Information Soluble in alcohol,chloroform,ether,and diethyl ether. Insoluble in water.
Formula Weight 167.98
Percent Purity ≥97%
Chemical Name or Material Allyl iodide, Stabilized with copper
Show More Show Less
Hazard Category H225-H300-H314-H335
Hazard Statement GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALLYL IODIDE
EINECSNumber 209-130-4
RTECSNumber UD0450000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only