Learn More
Allyl ethyl sulfide, 97%
CAS: 5296-62-8 | C5H10S | 102.20 g/mol
$183.59 - $183.59
Chemical Identifiers
| CAS | 5296-62-8 |
|---|---|
| Molecular Formula | C5H10S |
| Molecular Weight (g/mol) | 102.20 |
| MDL Number | MFCD00026979 |
| InChI Key | NOJXPGXFDASWEI-UHFFFAOYSA-N |
| Synonym | allyl ethyl sulfide, 4-thia-1-hexene, 1-propene-1-one, 3-ethylthio, allylethylsulfide, allyl ethylsulfide, allylethyl sulfide, allyl ethyl sulphide, 3-ethylthio-1-propene, acmc-1ao1o |
| PubChem CID | 123229 |
| IUPAC Name | 3-ethylsulfanylprop-1-ene |
| SMILES | CCSCC=C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1284706
|
Thermo Scientific Chemicals
L1284706 |
5 g |
Each for $183.59
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 5296-62-8 | |
| 102.20 | |
| NOJXPGXFDASWEI-UHFFFAOYSA-N | |
| 123229 | |
| CCSCC=C |
| C5H10S | |
| MFCD00026979 | |
| allyl ethyl sulfide, 4-thia-1-hexene, 1-propene-1-one, 3-ethylthio, allylethylsulfide, allyl ethylsulfide, allylethyl sulfide, allyl ethyl sulphide, 3-ethylthio-1-propene, acmc-1ao1o | |
| 3-ethylsulfanylprop-1-ene |
Specifications
| 5296-62-8 | |
| 115°C to 116°C | |
| Unpleasant | |
| 1.468 | |
| MFCD00026979 | |
| UN1993 | |
| allyl ethyl sulfide, 4-thia-1-hexene, 1-propene-1-one, 3-ethylthio, allylethylsulfide, allyl ethylsulfide, allylethyl sulfide, allyl ethyl sulphide, 3-ethylthio-1-propene, acmc-1ao1o | |
| CCSCC=C | |
| 102.20 | |
| 102.19 | |
| Allyl ethyl sulfide |
| 0.868 | |
| 18°C (64°F) | |
| C5H10S | |
| CH3CH2SCH2CH=CH2 | |
| 5 g | |
| 1736876 | |
| NOJXPGXFDASWEI-UHFFFAOYSA-N | |
| 3-ethylsulfanylprop-1-ene | |
| 123229 | |
| 97% |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only