missing translation for 'onlineSavingsMsg'
Learn More

Allyl Ethyl Carbonate 97.0+%, TCI America™

Supplier:  TCI America C128025ML

 View more versions of this product

Catalog No. C128025ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Allyl Ethyl Carbonate
Colorless
C6H10O3
25 mL
Carbonic Acid Allyl Ethyl Ester
CCOC(=O)OCC=C
130.143
130.14
Liquid
1469-70-1
149°C
MFCD00049113
3272
BGSFCOHRQUBESL-UHFFFAOYSA-N
ethyl prop-2-enyl carbonate
137020
≥97.0% (GC)

Chemical Identifiers

1469-70-1
130.143
BGSFCOHRQUBESL-UHFFFAOYSA-N
137020
CCOC(=O)OCC=C
C6H10O3
MFCD00049113
Carbonic Acid Allyl Ethyl Ester
ethyl prop-2-enyl carbonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions