accessibility menu, dialog, popup

UserName

CAS RN 1469-70-1

IUPAC Name: ethyl prop-2-en-1-yl carbonate
Synonyms: ethyl prop-2-en-1-yl carbonate
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: BGSFCOHRQUBESL-UHFFFAOYSA-N
SMILES: CCOC(=O)OCC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results