missing translation for 'onlineSavingsMsg'
Learn More

Allyl Ethyl Carbonate 97.0+%, TCI America™

Catalog No. C12805ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12805ML 5 mL
C128025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C12805ML Supplier TCI America Supplier No. C12805ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1469-70-1
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00049113
InChI Key BGSFCOHRQUBESL-UHFFFAOYSA-N
Synonym Carbonic Acid Allyl Ethyl Ester
PubChem CID 137020
IUPAC Name ethyl prop-2-enyl carbonate
SMILES CCOC(=O)OCC=C

Specifications

Color Colorless
Boiling Point 149°C
Quantity 5 mL
UN Number 3272
Formula Weight 130.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Ethyl Carbonate