missing translation for 'onlineSavingsMsg'
Learn More

Allyl Cyanoacetate 97.0+%, TCI America™

Catalog No. p-7124754
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C103925G 25 g
C1039500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C103925G Supplier TCI America Supplier No. C103925G

Chemical Identifiers

CAS 13361-32-5
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.13
MDL Number MFCD00013817
InChI Key WXKCRCGKCOKJEF-UHFFFAOYSA-N
Synonym Cyanoacetic Acid Allyl Ester
PubChem CID 25919
IUPAC Name prop-2-en-1-yl 2-cyanoacetate
SMILES C=CCOC(=O)CC#N

Specifications

Color Yellow
Boiling Point 101°C
Quantity 25 g
UN Number 3276
Formula Weight 125.13
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Cyanoacetate
EINECSNumber (2)-3241
RTECSNumber AG3690000
TSCA Yes