accessibility menu, dialog, popup

UserName

H2COON

CAS RN 13361-32-5

IUPAC Name: prop-2-en-1-yl 2-cyanoacetate
Synonyms: prop-2-en-1-yl 2-cyanoacetate
Molecular Weight (g/mol): 125.13
Molecular Formula: C6H7NO2
InChi Key: WXKCRCGKCOKJEF-UHFFFAOYSA-N
SMILES: C=CCOC(=O)CC#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results