missing translation for 'onlineSavingsMsg'
Learn More

Allyl Cyanide 92.0+%, TCI America™

Catalog No. A022725ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A022725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A022725ML Supplier TCI America Supplier No. A022725ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109-75-1
Molecular Formula C4H5N
Molecular Weight (g/mol) 67.091
MDL Number MFCD00001962
InChI Key SJNALLRHIVGIBI-UHFFFAOYSA-N
Synonym 3-butenenitrile, allyl cyanide, 3-butenylnitrile, 3-butenonitrile, allylnitrile, vinylacetonitrile, 3-cyanopropene, 3-cyano-1-propene, 1-butene-4-nitrile, propene-3-nitrile
PubChem CID 8009
IUPAC Name but-3-enenitrile
SMILES C=CCC#N

Specifications

Color Yellow
Boiling Point 118°C
Quantity 25 mL
UN Number 1992
Formula Weight 67.09
Percent Purity ≥92.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Cyanide
RTECSNumber EM8050000
TSCA Yes