accessibility menu, dialog, popup

UserName

CH2N

CAS RN 109-75-1

IUPAC Name: but-3-enenitrile
Synonyms: allyl cyanide
Molecular Weight (g/mol): 67.09
Molecular Formula: C4H5N
InChi Key: SJNALLRHIVGIBI-UHFFFAOYSA-N
SMILES: C=CCC#N
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)