missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allyl(chloromethyl)dimethylsilane 95.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Quantity | 5 g |
| UN Number | 1993 |
| Formula Weight | 148.71 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Allyl(chloromethyl)dimethylsilane |
Safety and Handling
| TSCA | No |