CAS RN 33558-75-7
IUPAC Name:
(chloromethyl)dimethyl(prop-2-en-1-yl)silane
Synonyms:
(chloromethyl)dimethylprop-2-en-1-ylsilane
Molecular Weight (g/mol):
148.71
Molecular Formula:
C6H13ClSi
InChi Key:
UHHLOVCFFWGSMO-UHFFFAOYSA-N
SMILES:
C[Si](C)(CCl)CC=C
1
–
1
of
1
results
1
Allyl(chloromethyl)dimethylsilane 95.0+%, TCI America™
CAS: 33558-75-7 Molecular Formula: C6H13ClSi Molecular Weight (g/mol): 148.705 MDL Number: MFCD00012045 InChI Key: UHHLOVCFFWGSMO-UHFFFAOYSA-N Synonym: 3-Chloromethyldimethylsilyl-1-propene PubChem CID: 556526 IUPAC Name: chloromethyl-dimethyl-prop-2-enylsilane SMILES: C[Si](C)(CC=C)CCl