missing translation for 'onlineSavingsMsg'
Learn More

Allyl Chloride 98.0+%, TCI America™

Catalog No. p-7124324
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
C027425ML 25 mL
C0274500ML 500 mL
2 options

Catalog No. C027425ML

Supplier: TCI America C027425ML

Only null left

Chemical Identifiers

CAS 107-05-1
Molecular Formula C3H5Cl
Molecular Weight (g/mol) 76.523
MDL Number MFCD00000984
InChI Key OSDWBNJEKMUWAV-UHFFFAOYSA-N
Synonym allyl chloride, 3-chloro-1-propene, 1-propene, 3-chloro, 3-chloropropylene, 3-chloropropene, chlorallylene, allylchlorid, 2-propenyl chloride, 1-chloro-2-propene, allylchloride
PubChem CID 7850
ChEBI CHEBI:82379
IUPAC Name 3-chloroprop-1-ene
SMILES C=CCCl

Specifications

Color Yellow
Boiling Point 45°C
Quantity 25 mL
UN Number 1100
Formula Weight 76.52
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Chloride
EINECSNumber (2)-0123
RTECSNumber UC7350000
TSCA Yes
Recommended Storage Refrigerator