missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Troemner™ Alloy 8 Metric Precision Weight Sets, Class 4 with NVLAP Certificate
Excellent for high-precision calibrations and reference standards
$2708.50 - $5171.76
Specifications
| Product Type | Medium Metric Analytical Weight Set |
|---|---|
| Certifications/Compliance | NVLAP, ISO9001 |
| Classification | ASTM Class 4 |
| Material | Stainless Steel |
| Catalog Number | Mfr. No. | Weight (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Weight (Metric) | Price | Quantity | |||||
|
0221550BC
|
Troemner™
30391240 |
10 mg to 100 g |
Each for $4,395.00
|
|
|||||
|
0221551CC
|
Troemner™
80850265 |
1 mg to 100 g |
Each for $5,171.76
|
|
|||||
|
0221550AC
|
Troemner™
30391236 |
10 mg to 50 g |
Each for $4,283.18
|
|
|||||
|
0221551BC
|
Troemner™
30391242 |
1 to 100 g |
Each for $2,708.50
|
|
|||||
Description
Designed for your most demanding weighing applications. Analytical Precision Weights meet or exceed specifications as defined in the ANSI/ASTM E617 standard.
- Low magnetic susceptibility, uniform consistency and high resistance to corrosion
- Used in extreme stable environments
- 1 to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
- Weight sets 1mg-500g are provided in a high quality polycarbonate set case
- Weight sets where the largest weight is 1kg and above are supplied in a felt line oak case
- Each polycarbonate case is placed in its own individual compartment
Certifications
A NIST/NVLAP™ or Traceable weight calibration certificate may be purchased to meet all of your ISO™, GLP, GMP, FDA, or nuclear requirements
Specifications
| Medium Metric Analytical Weight Set | |
| NVLAP, ISO9001 | |
| ASTM Class 4 | |
| Stainless Steel |