Learn More
Troemner™ Alloy 8 Metric Precision Weight Sets, Class 1
Excellent for high-precision calibrations and reference standards
Includes:
Forceps
Description
Manufactured from a specially developed austenitic stainless-stee—Alloy 8—available exclusively from Troemner. Weights 1g and larger are manufactured using Alloy 8, which possesses closely controlled density, extremely low magnetic susceptibility, excellent stability and resistance to corrosion, assuring a very stable mass over time. Other weights are stainless-steel.
- Each fractional weight is embossed with its value
- Weights from 1 to 30mg are aluminum and flat, with one side turned up to form a handle for non-slip gripping with forceps
- Weights from 50 to 500mg are stainless steel and flat with one side turned up to form a handle
- Gram and kilogram weights are manufactured using Alloy 8 and come in cylindrical shapes with knobs on top for easy handling
- Weight sets come in a 5-2-2-1 series, with two weights of any included mg or g value beginning with 2 (2mg, 20mg, etc.) and one weight for all other inclusive values
- Certificate of calibration is available; contact your Fisher Customer Service Representative
Specifications
| Medium Metric Analytical Weight Set | |
| Stainless Steel | |
| 100 g, 50 g, (2)20 g, 10 g, 5g, (2)2 g, 1 g | |
| 9/Set |
| ASTM Class 1 | |
| 1 to 100 g | |
| Cylindrical weight: 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, Polycarbonate case | |
| Multiple |