missing translation for 'onlineSavingsMsg'
Learn More

Allopurinol Impurity C, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022939
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022939 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022939 Supplier Merck Emd Millipore Supplier No. BP843

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

Chemical Name or Material Pharmacopeia & Metrological Institutes Standards
CAS 1346604-13-4
Flash Point Not Applicable
Molecular Formula C6H6N6O
Linear Formula C6H6N6O
Quantity 10 mg
Synonym 3-(4 H-1,2,4-Triazol-4-yl)-1 H-pyrazole-4-carboxamide; 5-(4 H-1,2,4-Triazol-4-yl)-1 H-pyrazole-4-carboxamide
Formula Weight 178.15
Grade BP