missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Allopurinol, 98%

Catalog No. AC121830050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC121830050 5 g Glass bottle
AC121830250 25 g Glass bottle
AC121831000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC121830050 Supplier Thermo Scientific Chemicals Supplier No. 121830050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 315-30-0 | C5H4N4O | 136.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 315-30-0
Molecular Formula C5H4N4O
Molecular Weight (g/mol) 136.11
MDL Number MFCD00599413
InChI Key OFCNXPDARWKPPY-UHFFFAOYSA-N
Synonym allopurinol, 1h-pyrazolo 3,4-d pyrimidin-4-ol, zyloprim, zyloric, lopurin, atisuril, bleminol, caplenal, uripurinol, embarin
PubChem CID 2094
ChEBI CHEBI:40279
IUPAC Name 1,2-dihydropyrazolo[3,4-d]pyrimidin-4-one
SMILES O=C1N=CN=C2NNC=C12

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 384.0°C
Color White
Loss on Drying 0.5 % (1 g, 105°C) max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 26,IV,2076
Merck Index 15,271
Solubility Information Solubility in water: 0.35g/L (25°C). Other solubilities: soluble in diluted alkalies and acids,practically insoluble in most common organic,solvents
Formula Weight 136.11
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Allopurinol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Toxic if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or ras
EINECSNumber 206-250-9

RUO – Research Use Only