missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Allopregnanolone ELISA Kit

Catalog No. EIAALLO
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EIAALLO 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EIAALLO Supplier Invitrogen™ Supplier No. EIAALLO

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ELISA

The Allopregnanolone Competitive ELISA quantitates allopregnanolone in serum, plasma, urine, saliva, dried fecal extracts or cell culture medium.∣Principle of the method: The Progesterone Metabolites Competitive ELISA research-use-only kit is designed to quantitatively measure allopregnanolone independent of species. A allopregnanolone standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with anti-rabbit IgG antibodies. Anti-rabbit allopregnanolone antibodies and allopregnanolone-peroxidase conjugate is added to the wells. The allopregnanolone-peroxidase conjugate and any allopregnanolone in the sample will compete to bind to the anti-rabbit allopregnanolone antibodies. After incubation, the plate is washed and substrate is added. The substrate reacts with the bound allopregnanolone-peroxidase conjugate. After a 30 min incubation, the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader at 450 nm. The intensity of the generated color corresponds inversely to the amount of allopregnanolone present in the sample.∣Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Specifications

Accession Number 3 alpha,5 alpha-tetrahydroprogesterone; 3 alpha-hydroxy-5 alpha-pregnan-20-one; allopregnanolone; Brexanolone; C21H34O2; Zulresso
Assay Range 156.25-10000 pg/mL
Assay Sensitivity 129.7 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Fecal Extract, Plasma, Serum, Supernatant, Urine
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 3alpha,5alpha-tetrahydroprogesterone; 3alpha-hydroxy-5alpha-pregnan-20-one; allopregnanolone; Brexanolone; C21H34O2; Zulresso3alpha,5alpha-tetrahydroprogesterone; 3alpha-hydroxy-5alpha-pregnan-20-one; allopregnanolone; Brexanolone; C21H34O2; Zulresso;
Interassay CV 8%
Intraassay CV 7%
Kit Contents Anti-IgG coated 96 well plate, Standard, Assay Buffer Concentrate, Capture Antibody, HRP-conjugated Antigen, Wash Buffer Concentrate, TMB Substrate, Stop Solution, Plate Sealer, Standard, Assay Buffer Concentrate, Capture Antibody, HRP-conjugated Antigen, Wash Buffer Concentrate, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Fecal Extract 50 μL, Plasma 50 μL, Serum 50 μL, Supernatant 50 μL, Urine 12.5 μL
Shipping Condition Wet or Dry Ice
Storage Requirements -20°C
Target Allopregnanolone
Test Time 45 min
Total Assay Time 2 hr 30 min
Show More Show Less