missing translation for 'onlineSavingsMsg'
Learn More

Allethrin, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC S4240

Catalog No. S4240

Only null left
Explore available promotions

Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Allethrin584-79-21000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Allethrin
584-79-2
C19H26O3
Certified Reference Material
CC1=C(C(=O)CC1OC(=O)C2C(C2(C)C)C=C(C)C)CC=C
(2-methyl-4-oxo-3-prop-2-enylcyclopent-2-en-1-yl) 2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylate
1 mL
CHEBI:34572
2mL amber prescored glass ampule
Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
ZCVAOQKBXKSDMS-UHFFFAOYSA-N
Methanol-P and T
302.414
11442
1000μg/mL

Chemical Identifiers

584-79-2
302.414
11442
(2-methyl-4-oxo-3-prop-2-enylcyclopent-2-en-1-yl) 2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylate
C19H26O3
ZCVAOQKBXKSDMS-UHFFFAOYSA-N
CHEBI:34572
CC1=C(C(=O)CC1OC(=O)C2C(C2(C)C)C=C(C)C)CC=C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Highly flammable liquid and vapor. Toxic if inhaled. Causes damage to organs.

DOTInformation : Nonhazardous

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only