Learn More
Allegro™ Respirator Cleaning Pads
Generously sized pads extend wear between complete cleanings.
Supplier: Allegro™ CAN3001
Description
- Designed specifically for use on rubber respirator facepieces and other personal safety equipment
- Available with 70% isopropanol or alcohol-free (with benzalkonium chloride) to clean even the most sensitive rubber facepiece
- Individually foil packed in quantities of 50/Pk. or 100/Pk.
Specifications
| Respirator Wipes | |
| Alcohol-free | |
| 100/Pk. | |
| 8 in. | |
| 5 in. |
| 3001 | |
| Respirator facepiece | |
| 8 x 5 in. (13 x 20 cm) | |
| 13 cm | |
| 20 cm |