Learn More
Allantoin, 98%
CAS: 97-59-6 | C4H6N4O3 | 158.117 g/mol
Supplier: Thermo Scientific Chemicals A155710B
Description
Allantoin is a useful biomarker for oxidative stress. It is a reactant for Synthesis of sulofenur analogs for use as antitumor agents. It is used as a cosmetic agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAllantoin is a useful biomarker for oxidative stress. It is a reactant for Synthesis of sulofenur analogs for use as antitumor agents. It is used as a cosmetic agent.
Solubility
Slightly soluble in water. Freely soluble in alkalis
Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
Specifications
| Allantoin | |
| ∼230°C (decomposition) | |
| MFCD00005260 | |
| 102364 | |
| allantoin, 5-ureidohydantoin, glyoxyldiureide, 1-2,5-dioxoimidazolidin-4-yl urea, glyoxyldiureid, allantol, cordianine, sebical, alantan, avc/dienestrolcream | |
| POJWUDADGALRAB-UHFFFAOYSA-N | |
| (2,5-dioxoimidazolidin-4-yl)urea | |
| 204 | |
| 158.12 |
| 97-59-6 | |
| C4H6N4O3 | |
| 1000 g | |
| 14,258 | |
| Slightly soluble in water. Freely soluble in alkalis | |
| C1(C(=O)NC(=O)N1)NC(=O)N | |
| 158.117 | |
| CHEBI:15676 | |
| 98% |
Chemical Identifiers
| 97-59-6 | |
| 158.117 | |
| POJWUDADGALRAB-UHFFFAOYSA-N | |
| 204 | |
| (2,5-dioxoimidazolidin-4-yl)urea |
| C4H6N4O3 | |
| MFCD00005260 | |
| allantoin, 5-ureidohydantoin, glyoxyldiureide, 1-2,5-dioxoimidazolidin-4-yl urea, glyoxyldiureid, allantol, cordianine, sebical, alantan, avc/dienestrolcream | |
| CHEBI:15676 | |
| C1(C(=O)NC(=O)N1)NC(=O)N |
Safety and Handling
EINECSNumber : 202-592-8
RTECSNumber : YT1600000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only