missing translation for 'onlineSavingsMsg'
Learn More

Allantoin, 98%

Catalog No. p-7022684
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1557130 250 g
AAA155710B 1000 g
AAA155710I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1557130 Supplier Thermo Scientific Chemicals Supplier No. A1557130

CAS: 97-59-6 | C4H6N4O3 | 158.117 g/mol

Allantoin is a useful biomarker for oxidative stress. It is a reactant for Synthesis of sulofenur analogs for use as antitumor agents. It is used as a cosmetic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allantoin is a useful biomarker for oxidative stress. It is a reactant for Synthesis of sulofenur analogs for use as antitumor agents. It is used as a cosmetic agent.

Solubility
Slightly soluble in water. Freely soluble in alkalis

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.

Chemical Identifiers

CAS 97-59-6
Molecular Formula C4H6N4O3
Molecular Weight (g/mol) 158.117
MDL Number MFCD00005260
InChI Key POJWUDADGALRAB-UHFFFAOYSA-N
Synonym allantoin, 5-ureidohydantoin, glyoxyldiureide, 1-2,5-dioxoimidazolidin-4-yl urea, glyoxyldiureid, allantol, cordianine, sebical, alantan, avc/dienestrolcream
PubChem CID 204
ChEBI CHEBI:15676
IUPAC Name (2,5-dioxoimidazolidin-4-yl)urea
SMILES C1(C(=O)NC(=O)N1)NC(=O)N

Specifications

Melting Point ∼230°C (decomposition)
Quantity 250 g
Beilstein 102364
Merck Index 14,258
Solubility Information Slightly soluble in water. Freely soluble in alkalis
Formula Weight 158.12
Percent Purity 98%
Chemical Name or Material Allantoin
EINECSNumber 202-592-8
RTECSNumber YT1600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only