Learn More
Brady™ All-weather Vinyl Labels
Make labels that withstand up to 10 years of outdoor abuse with this low-shrink vinyl label material (B-595). Print almost anywhere with a durable Brady™ handheld label maker.
Supplier:  Brady™ M4C750595CLBK
 View more versions of this product
Catalog No. 17012161
Description
Description
  • Ideal for labeling pipes, equipment, shelves and other industrial surfaces
  • Designed to withstand prolonged exposure to UV light, chemicals, greases and oils, salt spray, cleaners and detergents, and grime
  • Made with high-tack, permanent acrylic adhesive for use on rough, highly-textured, dirty and smooth surfaces alike, especially in cold temperatures
  • Weather-resistant labels designed to last up to 10 years in an outdoor environment
Specifications
Chemical Identifiers
Not Autoclavable
Permanent Acrylic
Gloss
Vinyl
Non-sterile
-40°C
-40°F to 180°F
0.004 in
0.5 in.
BS 5609:1986 Section 2 and section 3 Compliant with Directive 2011/65/EU and 2015/863
None
Continuous
Thermal Transfer
Component and equipment labelling;Electrical labelling;Facility identification;Floor marking;Lean and 5S labelling;Pipe and valve marking;Warehouse marking
1
-40°C to 82°C
0.102 mm
Specifications
Not Autoclavable
Permanent Acrylic
Transparent, Clear
Continuous
Top
Label
Component and equipment labelling;Electrical labelling;Facility identification;Floor marking;Lean and 5S labelling;Pipe and valve marking;Warehouse marking
-40°C
1 Label(s) / Box
19.05 mm (W) x 6.10 m (L)
-40°C to 82°C
0.102 mm
20 ft.
0.75 in
19.05 mm
M4C-750-595-CL-BK
BS 5609:1986 Section 2 and section 3 Compliant with Directive 2011/65/EU and 2015/863
None
Gloss
Vinyl
Thermal Transfer
Non-sterile
BMP41, BMP51, BMP53, M511, M410, M510
M4C-750-595-CL-BK
1
-40°F to 180°F
0.004 in
19.05 mm (W) x 6.10 m (L)
6.10 m
19.05 mm
0.5 in.
Product Suggestions
Product Suggestions