Learn More
Keystone Cleanroom Products All Tyvek Bag, With Steam Indicator Dot
All 1073B Tyvek Autoclave Bags are the best option for heavy equipment parts, or those with sharp edges and corners that will puncture inferior wrapping materials.
$2,623.26 - $4,958.41
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Certifications/Compliance | CoC |
| Closure Type | Heat Seal |
| Color | White |
| Disposable | Single-use |
| Catalog Number | Mfr. No. | Length (English) | Width (English) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Length (English) | Width (English) | Price | Quantity & Availability | ||||
|
19999160
|
Keystone Cleanroom Products
60114TW1073BSD |
14 in. | 1 in. |
Case of 500 Each for $3,669.52
|
|
||||
|
19999170
|
Keystone Cleanroom Products
60304TW1073BSD |
4 in. | 3 in. |
Case of 500 Each for $3,942.47
|
|
||||
|
19999171
|
Keystone Cleanroom Products
60306TW1073BSD |
6 in. | 3 in. |
Case of 500 Each for $4,169.90
|
|
||||
|
19999174
|
Keystone Cleanroom Products
60816TW1073BSD |
16 in. | 8 in. |
Case of 250 Each for $3,487.56
|
|
||||
|
19999175
|
Keystone Cleanroom Products
61212TW1073BSD |
12 in. | 12 in. |
Case of 250 Each for $3,199.46
|
|
||||
|
19999176
|
Keystone Cleanroom Products
61222TW1073BSD |
22 in. | 12 in. |
Case of 250 Each for $4,215.40
|
|
||||
|
19999177
|
Keystone Cleanroom Products
61224TW1073BSD |
24 in. | 12 in. |
Case of 250 Each for $4,958.41
|
|
||||
|
19999178
|
Keystone Cleanroom Products
61518TW1073BSD |
18 in. | 15 in. |
Case of 250 Each for $4,215.40
|
|
||||
|
19999179
|
Keystone Cleanroom Products
61822TW1073BSD |
22 in. | 18 in. |
Case of 125 Each for $2,623.26
|
|
||||
|
19999180
|
Keystone Cleanroom Products
61826TW1073BSD |
26 in. | 18 in. |
Case of 125 Each for $2,623.26
|
|
||||
|
19999181
|
Keystone Cleanroom Products
62424TW1073BSD |
24 in. | 24 in. |
Case of 125 Each for $2,987.17
|
|
||||
|
19999182
|
Keystone Cleanroom Products
62630TW1073BSD |
30 in. | 26 in. |
Case of 125 Each for $2,623.26
|
|
||||
|
19999183
|
Keystone Cleanroom Products
63030TW1073BSD |
30 in. | 30 in. |
Case of 125 Each for $3,563.39
|
|
||||
|
19999184
|
Keystone Cleanroom Products
63636TW1073BSD |
36 in. | 36 in. |
Case of 125 Each for $3,987.96
|
|
||||
|
19999185
|
Keystone Cleanroom Products
63642TW1073BSD |
42 in. | 36 in. |
Case of 125 Each for $4,306.38
|
|
||||
|
19999186
|
Keystone Cleanroom Products
63648TW1073BSD |
48 in. | 36 in. |
Case of 125 Each for $4,912.92
|
|
||||
|
19999187
|
Keystone Cleanroom Products
64048TW1073BSD |
48 in. | 40 in. |
Case of 125 Each for $4,912.92
|
|
||||
Description
- Tyvek™ 1073B bags are ideal for steam sterilization of pharmaceutical components such as vial and syringe stoppers and caps as well as heavy equipment parts with sharp edges.
- Manufactured and packed in an ISO 6 Certified cleanroom
- Steam sterilisation indicator - autoclavable at 121 to 125 °C
- Tear and puncture resistant, hydrophobic and lint-free
- 100% medical grade Tyvek™ 1073B allows fast steam penetration
- Excellent impact resistance with exceptional microbial barrier
- Consistently strong factory seals
- 100% medical and pharmaceutical grade Tyvek™ 1073B autoclave bags are ideal for steam sterilization of pharmaceutical components such as vial and syringe stoppers and caps.
- With consistently strong factory seals, these are a good choice for heavy or pharmaceutical change parts with sharp corners.
- All Tyvek construction allows for a greater number of bags per sterilisation load; increasing efficiencies and cost reduction.
Specifications
| Autoclavable | |
| Heat Seal | |
| Single-use | |
| Non-sterile | |
| Autoclave Bag |
| CoC | |
| White | |
| Tyvek | |
| Yes |