missing translation for 'onlineSavingsMsg'
Learn More

all-trans-N-(4-Hydroxyphenyl)retinamide 98.0+%, TCI America™

Catalog No. p-7131058
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H146410MG 10 mg
H1464100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H146410MG Supplier TCI America Supplier No. H146410MG
Only null left

Chemical Identifiers

CAS 65646-68-6
Molecular Formula C26H33NO2
Molecular Weight (g/mol) 391.555
MDL Number MFCD00792674
InChI Key AKJHMTWEGVYYSE-FXILSDISSA-N
Synonym fenretinide, n-4-hydroxyphenyl retinamide, 4-hpr, 4-hydroxyphenylretinamide, 4-hydroxyphenyl retinamide, retinoic acid p-hydroxyanilide, all-trans-4'-hydroxyretinanilide, fenretinida, fenretinidum, fenretinidum latin
PubChem CID 5288209
ChEBI CHEBI:42588
IUPAC Name (2E,4E,6E,8E)-N-(4-hydroxyphenyl)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenamide
SMILES CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CC(=O)NC2=CC=C(C=C2)O)C)C

Specifications

Melting Point 172°C
Quantity 10 mg
Formula Weight 391.56
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material all-trans-N-(4-Hydroxyphenyl)retinamide
RTECSNumber VH6420000
TSCA No
Recommended Storage Freezer (-20 °C)