missing translation for 'onlineSavingsMsg'
Learn More

All-rac-a-Tocopheryl Acetate, FCC, 96-102%, Spectrum™ Chemical

V1048, 7695-91-2, C31H52O3

$883.73 - $5,721.97

Chemical Identifiers

CAS 7695-91-2
Molecular Formula C31H52O3
Molecular Weight (g/mol) 472.75
InChI Key ZAKOWWREFLAJOT-CEFNRUSXSA-N
IUPAC Name (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-yl acetate
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(OC(C)=O)=C(C)C(C)=C2O1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18600413
View Documents Promotion Details
N/A 100 mL Amber Glass Bottle
Each for $883.73
Only null left
 
18600414
View Documents Promotion Details
N/A 1 L Amber Glass Bottle
Each for $5,721.97
Only null left
 
Description

Description

Spectrum™ Chemical All-rac-a-Tocopheryl Acetate, FCC is the synthetic form of Vitamin E. Alpha-tocopheryl acetate is more resistant to oxidation during storage than unesterified tocopherols. The bioavailability of alpha-tocopherol from alpha-tocopheryl acetate is roughly equivalent to that of free alpha-tocopherol. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

7695-91-2
472.75
(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-yl acetate
C31H52O3
ZAKOWWREFLAJOT-CEFNRUSXSA-N
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(OC(C)=O)=C(C)C(C)=C2O1

Specifications

7695-91-2
443°C
ZAKOWWREFLAJOT-CEFNRUSXSA-N
(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-yl acetate
96 to 102%
Liquid
100%
C31H52O3
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(OC(C)=O)=C(C)C(C)=C2O1
472.75
FCC
All-rac-a-Tocopheryl Acetate
Videos
SDS
Documents

Documents