missing translation for 'onlineSavingsMsg'
Learn More

Alkalinity, CaCO3 1,000 mg/L Calibration Standard, MilliporeSigma™ Supelco™

Catalog No. 111004891
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004891 100 mL
111016358 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111004891 Supplier Merck Emd Millipore Supplier No. ALK1000100ML

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Flavors and Fragrances, Food and Beverages and Personal Care

Specifications

Type Certified Reference Material
Quantity 100 mL
Grade Certified Reference Material