missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Solution (Sodium Hydroxide and Potassium Sodium Tartrate), Ricca Chemical Company
Specifications
Specifications
| Chemical Name or Material | Alkaline Solution |
| Color | Colorless |
| CAS | 1310-73-2, 5381-59-5, 7732-18-5 |
| Boiling Point | ~100°C |
| Grade | Laboratory |
| pH | >12 |
| Quantity | 4 L |
| Sterility | Non-sterile |