missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Pyrogallol Reagent, Orsat, Ricca Chemical
Specifications
Specifications
| CAS Min % | 4.0, 25.0, 65.0 |
| CAS Max % | 6.0, 35.0, 75.0 |
| Packaging | Amber Poly Bottle |
| Quantity | 1 L |
| Chemical Name or Material | Alkaline Pyrogallol Reagent |