missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Iodide Azide Solution I, For Dissolved Oxygen (Saturated and Less than Saturated Samples), Certified, LabChem™
$119.04 - $743.79
Specifications
| Color | Clear, colorless |
|---|---|
| Format | Liquid |
| Storage Requirements | Room Temperature |
| Type | Alkaline Iodide Azide Solution I |
Specifications
| Clear, colorless | |
| Liquid | |
| Room Temperature | |
| Alkaline Iodide Azide Solution I |