missing translation for 'onlineSavingsMsg'
Learn More

Alkaline Iodide Azide Solution I, For Dissolved Oxygen (Saturated and Less than Saturated Samples), Certified, LabChem™

Catalog No. LC106701
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Poly
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC106701 500 mL Amber Poly
LC106702 1 L Amber Poly
LC106704 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC106701 Supplier LabChem Supplier No. LC106701

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Color Clear, colorless
Format Liquid
Packaging Amber Poly
Quantity 500 mL
Storage Requirements Room Temperature
Type Alkaline Iodide Azide Solution I