Learn More
Invitrogen™ Human ALK [F1174L], GST Tag, His Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Human ALK F1174L, internal fragment, amino acids L1066-S1437 (as in NCBI/Protein entry NP_004295.2), F1174L mutation, N-terminal GST-HIS6 fusion protein with a 3C cleavage site, expressed in Sf9 insect cells. , Validated in LanthaScreen™ Kinase Binding Assay, Protocols available for simple use and optimized performance, Multiple pack sizes available for convenient use Related Product: LanthaScreen™ Eu Kinase Binding Assay Protein Form: Recombinant
Specifications
Specifications
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM HEPES with 100 mM NaCl, 15 mM glutathione, 20% glycerol, 5 mM DTT and no preservative; pH 7.5 |
| Molecular Weight (g/mol) | 70.4 kDa |
| Name | Human ALK [F1174L], GST Tag, His Tag |
| Quantity | 10 μg |
| Common Name | ALK (F1174L Mutant) |
| Protein Length | 1066-1437 |
| Conjugate | Unconjugated |
| Species | Human |
| Show More |