missing translation for 'onlineSavingsMsg'
Learn More

Alizarin 95.0+%, TCI America™

Supplier:  TCI America D024225G

Catalog No. D024225G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Alizarin
290°C
C14H8O4
25 g
RGCKGOZRHPZPFP-UHFFFAOYSA-N
1,2-dihydroxyanthracene-9,10-dione
6293
240.21
Crystalline Powder
72-48-0
430°C
MFCD00001201
alizarin, alizarin red, mordant red 11, 1,2-dihydroxyanthraquinone, alizarin b, turkey red, 1,2-anthraquinonediol, alizarine, alizarina, alizarine red
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C=C3)O)O
240.214
CHEBI:16866
≥95.0% (HPLC)

Chemical Identifiers

72-48-0
240.214
RGCKGOZRHPZPFP-UHFFFAOYSA-N
6293
1,2-dihydroxyanthracene-9,10-dione
C14H8O4
MFCD00001201
alizarin, alizarin red, mordant red 11, 1,2-dihydroxyanthraquinone, alizarin b, turkey red, 1,2-anthraquinonediol, alizarine, alizarina, alizarine red
CHEBI:16866
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C=C3)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0704

RTECSNumber : CB6580000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions