accessibility menu, dialog, popup

UserName

HOOOOH

CAS RN 72-48-0

IUPAC Name: 1,2-dihydroxy-9,10-dihydroanthracene-9,10-dione
Synonyms: alizarin alizarine paste 20 percent bluish alizarine lake red 3P alizarine lake red 2P alizarine indicator alizarine red L alizarine red B alizarin B alizarine B alizarine red IP
Molecular Weight (g/mol): 240.21
Molecular Formula: C14H8O4
InChi Key: RGCKGOZRHPZPFP-UHFFFAOYSA-N
SMILES: OC1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1O
Percent Purity 
  • (1)
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (1)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (7)
  • (1)
  • (3)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)