missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alizarin Red S, Spectrum™ Chemical
AL200, 130-22-3, C14H7O7SNa·H2O
$437.68 - $1101.21
Chemical Identifiers
| CAS | 130-22-3 |
|---|---|
| Molecular Formula | C14H7NaO7S |
| Molecular Weight (g/mol) | 342.25 |
| InChI Key | HFVAFDPGUJEFBQ-UHFFFAOYSA-M |
| IUPAC Name | sodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulfonate |
| SMILES | [Na+].OC1=C(O)C2=C(C=C1S([O-])(=O)=O)C(=O)C1=CC=CC=C1C2=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18600395
|
Spectrum Chemical Mfg Cor
AL20010GM |
10 g | Amber Glass Bottle |
Each for $437.68
|
|
||||
|
18600396
|
Spectrum Chemical Mfg Cor
AL20025GM |
25 g | Amber Glass Bottle |
Each for $581.64
|
|
||||
|
18600397
|
Spectrum Chemical Mfg Cor
AL200100GM |
100 g | Amber Glass Bottle |
Each for $1,101.21
|
|
||||
Description
Spectrum™ Chemical Alizarin Red S , also known as 1,2-dihydroxyanthraquinone, can be used as a dye for textile fabrics. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Chemical Identifiers
| 130-22-3 | |
| 342.25 | |
| sodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulfonate |
| C14H7NaO7S | |
| HFVAFDPGUJEFBQ-UHFFFAOYSA-M | |
| [Na+].OC1=C(O)C2=C(C=C1S([O-])(=O)=O)C(=O)C1=CC=CC=C1C2=O |
Specifications
| 130-22-3 | |
| C14H7NaO7S | |
| [Na+].OC1=C(O)C2=C(C=C1S([O-])(=O)=O)C(=O)C1=CC=CC=C1C2=O | |
| 342.25 | |
| Amber Glass Bottle | |
| 10 g |
| 100% | |
| HFVAFDPGUJEFBQ-UHFFFAOYSA-M | |
| sodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulfonate | |
| Reagent | |
| Red/Brown to Dark Red | |
| Alizarin Red S |