missing translation for 'onlineSavingsMsg'
Learn More

Alizarin Red S, Spectrum™ Chemical

Catalog No. 18600397
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600397 100 g Amber Glass Bottle
18600395 10 g Amber Glass Bottle
18600396 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18600397 Supplier Spectrum Chemical Mfg Cor Supplier No. AL200100GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AL200, 130-22-3, C14H7O7SNa·H2O

Spectrum™ Chemical Alizarin Red S , also known as 1,2-dihydroxyanthraquinone, can be used as a dye for textile fabrics. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 130-22-3
Molecular Formula C14H7NaO7S
Molecular Weight (g/mol) 342.25
InChI Key HFVAFDPGUJEFBQ-UHFFFAOYSA-M
IUPAC Name sodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulfonate
SMILES [Na+].OC1=C(O)C2=C(C=C1S([O-])(=O)=O)C(=O)C1=CC=CC=C1C2=O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Color Red/Brown to Dark Red
Quantity 100 g
Grade Reagent
Chemical Name or Material Alizarin Red S