missing translation for 'onlineSavingsMsg'
Learn More

Alizarin Red S (Sodium Sulfonate), MP Biomedicals™

Alizarin red S is used in a biochemical assay to determine, quantitatively by colorimetry, the presence of calcific deposition by cells of an osteogenic lineage

Supplier:  MP Biomedicals Inc 0210037580

 View more versions of this product

Catalog No. ICN10037580


Only null left

Description

Description

Specifications

Specifications

Alizarin Red S
130-22-3
MFCD00013049
HFVAFDPGUJEFBQ-UHFFFAOYSA-M
sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate
3955344
342.3
100 g
Indicator: pH Range 3.7 - 5.2
C14H7NaO7S
Sodium Alizarinsulfonate, Alizarin sodium sulfonate
C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+]
342.253
CHEBI:87358
Orange
Powder

Chemical Identifiers

130-22-3
342.253
HFVAFDPGUJEFBQ-UHFFFAOYSA-M
3955344
sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate
C14H7NaO7S
MFCD00013049
Sodium Alizarinsulfonate, Alizarin sodium sulfonate
CHEBI:87358
C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 204-981-8

Recommended Storage : Store at Room Temperature (15-30°C).

SDS
Documents

Documents

Promotions

Promotions