missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Alizarin Red S sodium salt, 1% w/v aq. sol.

Transition interval: pH 4.6 (yellow) to pH 6.0 (red) | CAS: 130-22-3 | C14H7NaO7S | 342.253 g/mol

Supplier:  Thermo Scientific Chemicals 042746AE

 View more versions of this product

Catalog No. AA42746AE


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Alizarin Red S sodium salt
C14H7NaO7S
HFVAFDPGUJEFBQ-UHFFFAOYSA-M
sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate
3955344
342.27
Amber
Liquid
130-22-3
MFCD00013049
C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+]
342.253
CHEBI:87358
Odorless
100 mL

Chemical Identifiers

130-22-3
342.253
HFVAFDPGUJEFBQ-UHFFFAOYSA-M
CHEBI:87358
C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+]
C14H7NaO7S
MFCD00013049
3955344
sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only