missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Alizarin Red S sodium salt, 1% w/v aq. sol.

Catalog No. p-4824576
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42746AE 100 mL
AA42746AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42746AE Supplier Thermo Scientific Chemicals Supplier No. 042746AE

Transition interval: pH 4.6 (yellow) to pH 6.0 (red) | CAS: 130-22-3 | C14H7NaO7S | 342.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 130-22-3
Molecular Formula C14H7NaO7S
Molecular Weight (g/mol) 342.253
MDL Number MFCD00013049
InChI Key HFVAFDPGUJEFBQ-UHFFFAOYSA-M
PubChem CID 3955344
ChEBI CHEBI:87358
IUPAC Name sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate
SMILES C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+]

Specifications

Odor Odorless
Color Amber
Quantity 100 mL
Physical Form Liquid
Formula Weight 342.27
Chemical Name or Material Alizarin Red S sodium salt
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only