missing translation for 'onlineSavingsMsg'
Learn More

Alizarin Red S, 1% Aqueous, Certified, LabChem™

Supplier:  LabChem LC106002

 View more versions of this product

Catalog No. LC106002


Only null left

Description

Description

Specifications

Specifications

Alizarin Red S
130-22-3 , 7732-18-5
C14H7NaO7S
Soluble in water
HFVAFDPGUJEFBQ-UHFFFAOYSA-M
sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate
3955344
342.27
Poly Bottle
Carbon monoxide; Carbon dioxide; Nitrogen oxides
Red
Liquid
1% Aqueous
99,1
C14H7NaO7S
Passes Test
C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+]
342.253
CHEBI:87358
Certified
1g/mL
1g/mL
1 L

Chemical Identifiers

130-22-3
342.253
3955344
sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate
C14H7NaO7S
HFVAFDPGUJEFBQ-UHFFFAOYSA-M
CHEBI:87358
C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions