missing translation for 'onlineSavingsMsg'
Learn More

Alizarin Red S, 1% Aqueous, Certified, LabChem™

Catalog No. LC106007
Change view
Click to view available options
Quantity:
125 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC106007 125 mL
LC106001 500 mL
LC106002 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC106007 Supplier LabChem Supplier No. LC106007

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 130-22-3 , 7732-18-5
Molecular Formula C14H7NaO7S
Molecular Weight (g/mol) 342.253
InChI Key HFVAFDPGUJEFBQ-UHFFFAOYSA-M
PubChem CID 3955344
ChEBI CHEBI:87358
IUPAC Name sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate
SMILES C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+]

Specifications

CAS Max % 99, 1
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Linear Formula C14H7NaO7S
Decomposition Information Carbon monoxide; Carbon dioxide; Nitrogen oxides
Density 1g/mL
Color Red
Solubility Information Soluble in water
Quantity 125 mL
Physical Form Liquid
Formula Weight 342.27
Grade Certified
Chemical Name or Material Alizarin Red S, 1% Aqueous
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature