missing translation for 'onlineSavingsMsg'
Learn More

Alizarin, 94%

CAS: 72-48-0 | C14H8O4 | 240.214 g/mol

$109.10 - $1941.84

Chemical Identifiers

CAS 72-48-0
Molecular Formula C14H8O4
Molecular Weight (g/mol) 240.214
MDL Number MFCD00001201
InChI Key RGCKGOZRHPZPFP-UHFFFAOYSA-N
Synonym alizarin, alizarin red, mordant red 11, 1,2-dihydroxyanthraquinone, alizarin b, turkey red, 1,2-anthraquinonediol, alizarine, alizarina, alizarine red
PubChem CID 6293
ChEBI CHEBI:16866
IUPAC Name 1,2-dihydroxyanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C=C3)O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1440422
View Documents
Thermo Scientific Chemicals
A1440422
100 g
Each for $109.10
Only null left
 
AAA1440436
View Documents
Thermo Scientific Chemicals
A1440436
500 g
Each for $430.74
Only null left
 
AAA144040E
View Documents
Thermo Scientific Chemicals
A144040E
2500 g
Each for $1,941.84
Only null left
 
Description

Description

Alizarin has a major application in the manufacture of the madder lake pigments which is commonly called as Rose madder and Alizarin crimson. It is involved in the studies of bone growth, osteoporosis, bone marrow and calcium deposits in the vascular system. It is also used as a stain in calcite and aragonite to identify calcium carbonate, and synovial fluid to asses for basic calcium phosphate crystals. It is used in colorimetric measurements for quantification of amine extraction by model food simulants from epoxy polymer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Alizarin has a major application in the manufacture of the madder lake pigments which is commonly called as Rose madder and Alizarin crimson. It is involved in the studies of bone growth, osteoporosis, bone marrow and calcium deposits in the vascular system. It is also used as a stain in calcite and aragonite to identify calcium carbonate, and synovial fluid to asses for basic calcium phosphate crystals. It is used in colorimetric measurements for quantification of amine extraction by model food simulants from epoxy polymer.

Solubility
Soluble in hexane and chloroform. Slightly soluble in water.

Notes
Incompatible with alkalis and oxidizing agents.
Specifications

Chemical Identifiers

72-48-0
240.214
RGCKGOZRHPZPFP-UHFFFAOYSA-N
6293
1,2-dihydroxyanthracene-9,10-dione
C14H8O4
MFCD00001201
alizarin, alizarin red, mordant red 11, 1,2-dihydroxyanthraquinone, alizarin b, turkey red, 1,2-anthraquinonediol, alizarine, alizarina, alizarine red
CHEBI:16866
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C=C3)O)O

Specifications

72-48-0
∼430°C (sublimation)
MFCD00001201
1914037
alizarin, alizarin red, mordant red 11, 1,2-dihydroxyanthraquinone, alizarin b, turkey red, 1,2-anthraquinonediol, alizarine, alizarina, alizarine red
RGCKGOZRHPZPFP-UHFFFAOYSA-N
1,2-dihydroxyanthracene-9,10-dione
6293
240.21
Alizarin
286°C to 290°C
C14H8O4
100 g
14,251
Soluble in hexane and chloroform. Slightly soluble in water.
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C=C3)O)O
240.214
CHEBI:16866
94%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c

H302-H319

EINECSNumber : 200-782-5

RTECSNumber : CB6580000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only