missing translation for 'onlineSavingsMsg'
Learn More

Aliphatics Mix (C5-C12), Certified Reference Material, 2,000 μg/mL Each Component in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111017041
Change view
Click to view available options
Quantity:
1.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017041 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017041 Supplier Merck Emd Millipore Supplier No. UST1571.5ML

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Specifications

Concentration 2,000 μg/mL each component in methanol
Quantity 1.5 mL
Grade Certified Reference Material
Concentration or Composition (by Analyte or Components) Pentane; Hexane; Heptane; Octane; Nonane; Decane; Undecane; Dodecane