missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alimemazine Tartrate 98.0+%, TCI America™
$65.46 - $1143.56
Chemical Identifiers
| CAS | 4330-99-8 |
|---|---|
| Molecular Formula | C40H50N4O6S2 |
| Molecular Weight (g/mol) | 746.982 |
| MDL Number | MFCD00242594 |
| InChI Key | AJZJIYUOOJLBAU-RNKHSWPKSA-N |
| Synonym | Trimeprazine Tartrate, 10-[3-(Dimethylamino)-2-methylpropyl]phenothiazine Tartrate |
| PubChem CID | 45358050 |
| IUPAC Name | (2S,3S)-2,3-dihydroxybutanedioic acid;N,N,2-trimethyl-3-phenothiazin-10-ylpropan-1-amine |
| SMILES | CC(CN1C2=CC=CC=C2SC3=CC=CC=C31)CN(C)C.CC(CN1C2=CC=CC=C2SC3=CC=CC=C31)CN(C)C.C(C(C(=O)O)O)(C(=O)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A2813100MG
|
TCI America
A2813100MG |
100 mg |
Each for $65.46
|
|
|||||
|
A28131G
|
TCI America
A28131G |
1 g |
Each for $381.18
|
|
|||||
|
A28135G
|
TCI America
A28135G |
5 g |
Each for $1,143.56
|
|
|||||
Chemical Identifiers
| 4330-99-8 | |
| 746.982 | |
| AJZJIYUOOJLBAU-RNKHSWPKSA-N | |
| 45358050 | |
| CC(CN1C2=CC=CC=C2SC3=CC=CC=C31)CN(C)C.CC(CN1C2=CC=CC=C2SC3=CC=CC=C31)CN(C)C.C(C(C(=O)O)O)(C(=O)O)O |
| C40H50N4O6S2 | |
| MFCD00242594 | |
| Trimeprazine Tartrate, 10-[3-(Dimethylamino)-2-methylpropyl]phenothiazine Tartrate | |
| (2S,3S)-2,3-dihydroxybutanedioic acid;N,N,2-trimethyl-3-phenothiazin-10-ylpropan-1-amine |
Specifications
| 4330-99-8 | |
| White-Yellow | |
| MFCD00242594 | |
| 2811 | |
| AJZJIYUOOJLBAU-RNKHSWPKSA-N | |
| (2S,3S)-2,3-dihydroxybutanedioic acid;N,N,2-trimethyl-3-phenothiazin-10-ylpropan-1-amine | |
| 45358050 | |
| ≥98.0% (HPLC,T) | |
| Alimemazine Tartrate |
| 161°C | |
| C40H50N4O6S2 | |
| 100 mg | |
| Trimeprazine Tartrate, 10-[3-(Dimethylamino)-2-methylpropyl]phenothiazine Tartrate | |
| CC(CN1C2=CC=CC=C2SC3=CC=CC=C31)CN(C)C.CC(CN1C2=CC=CC=C2SC3=CC=CC=C31)CN(C)C.C(C(C(=O)O)O)(C(=O)O)O | |
| 746.982 | |
| 746.98 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : SO6475000
TSCA : No