missing translation for 'onlineSavingsMsg'
Learn More

Alimemazine Tartrate 98.0+%, TCI America™

Supplier:  TCI America A28131G

 View more versions of this product

Catalog No. A28131G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Alimemazine Tartrate
161°C
C40H50N4O6S2
1 g
Trimeprazine Tartrate, 10-[3-(Dimethylamino)-2-methylpropyl]phenothiazine Tartrate
CC(CN1C2=CC=CC=C2SC3=CC=CC=C31)CN(C)C.CC(CN1C2=CC=CC=C2SC3=CC=CC=C31)CN(C)C.C(C(C(=O)O)O)(C(=O)O)O
746.982
746.98
Crystalline Powder
4330-99-8
White-Yellow
MFCD00242594
2811
AJZJIYUOOJLBAU-RNKHSWPKSA-N
(2S,3S)-2,3-dihydroxybutanedioic acid;N,N,2-trimethyl-3-phenothiazin-10-ylpropan-1-amine
45358050
≥98.0% (HPLC,T)

Chemical Identifiers

4330-99-8
746.982
AJZJIYUOOJLBAU-RNKHSWPKSA-N
45358050
CC(CN1C2=CC=CC=C2SC3=CC=CC=C31)CN(C)C.CC(CN1C2=CC=CC=C2SC3=CC=CC=C31)CN(C)C.C(C(C(=O)O)O)(C(=O)O)O
C40H50N4O6S2
MFCD00242594
Trimeprazine Tartrate, 10-[3-(Dimethylamino)-2-methylpropyl]phenothiazine Tartrate
(2S,3S)-2,3-dihydroxybutanedioic acid;N,N,2-trimethyl-3-phenothiazin-10-ylpropan-1-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : SO6475000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions